Order Entry
United States
ContactUsLinkComponent
Cynomolgus Monkey Recombinant Siglec-15 (from HEK293 cells)
Cynomolgus Monkey Recombinant Siglec-15 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Cynomolgus Monkey Recombinant Siglec-15 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Tag sequence:
    Fc Tag
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized Cynomolgus Siglec-15, Fc Tag (Cat. No. SG5-C5253) at 5 μg/mL (100 μL/well) can bind Mouse Human chimeric MAb (5G12, Human IgG1) with a linear range of 0.4-6 ng/mL (QC tested). Measured by its binding ability in a functional ELISA. Immobilized Cynomolgus Siglec-15, Fc Tag (Cat. No. SG5-C5253) at 10 μg/mL (100 μL/well) on Diamond Protein A Protein, His Tag precoated (0.5 μg/well) plate, can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with a linear range of 0.078-1.25 μg/mL (Routinely tested). Loaded Cynomolgus Siglec-15, Fc Tag (Cat. No. SG5-C5253) on ProteinA Biosensor, can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with an affinity constant of 0.62 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    A0A2K5UY47-1
  • Protein Synonyms:
    Siglec-15|SIGLEC-15|Siglec15|CD33L3|CD33 antigen-like 3|sialic acid-binding Ig-like lectin 15
  • Protein/Peptide Name:
    Siglec-15
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    52.7 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together