Order Entry
United States
ContactUsLinkComponent
Cynomolgus Monkey Recombinant Latent TGF-beta 1 (from HEK293 cells)
Cynomolgus Monkey Recombinant Latent TGF-beta 1 (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
Cynomolgus Monkey Recombinant Latent TGF-beta 1 (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    A0A2K5TJB2-1 (C33S)
  • Protein Synonyms:
    CED|TGF-beta-1|TGFB|DPD1|Latent TGF-beta 1|Latent TGFB1|LAP|TGFB1
  • Protein/Peptide Name:
    Latent TGF-beta 1
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    43.1 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together