Order Entry
United States
ContactUsLinkComponent
Cynomolgus Monkey, Rhesus macaque Recombinant FcRn (FCGRT & B2M) (from HEK293 cells), Biotin
Cynomolgus Monkey, Rhesus macaque Recombinant FcRn (FCGRT & B2M) (from HEK293 cells), Biotin
Supplier:  ACROBIOSYSTEMS
Certificates
Cynomolgus Monkey, Rhesus macaque Recombinant FcRn (FCGRT & B2M) (from HEK293 cells), Biotin
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Biotin
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
    Rhesus monkey
  • Tag sequence:
    His, Avitag™
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Biotinylated Cynomolgus / Rhesus macaque FcRn Heterodimer Protein, His,Avitag (SPR verified) (Cat. No. FCM-C82W3) captured on Biotin CAP-Series S Sensor Chip can bind Herceptin® with an affinity constant of 0.119 μM as determined in a SPR assay (Biacore T200) (QC tested). Loaded Biotinylated Cynomolgus / Rhesus macaque FcRn Heterodimer Protein, His,Avitag (SPR verified)(Cat. No. FCM-C82W3) on SA Biosensor, can bind Cynomolgus Serum Albumin, His Tag(Cat. No. CSA-C52H4) with an affinity constant of 0.617 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    Q8SPV9-1 (FCGRT) & Q8SPW0-1 (B2M)
  • Protein Synonyms:
    FcRn|FCGRT & B2M
  • Protein/Peptide Name:
    FcRn (FCGRT & B2M)
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    34.1 kDa (FCGRT) & 11.6 kDa (B2M)
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together