Order Entry
United States
ContactUsLinkComponent
Cynomolgus Monkey, Rhesus macaque Recombinant CD47 (from HEK293 cells)
Cynomolgus Monkey, Rhesus macaque Recombinant CD47 (from HEK293 cells)
Catalog # 103790-186
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Cynomolgus Monkey, Rhesus macaque Recombinant CD47 (from HEK293 cells)
Catalog # 103790-186
Supplier:  ACROBIOSYSTEMS
Supplier Number:  CD7C52H11MG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
    Rhesus monkey
  • Size:
    1 mg
  • Tag sequence:
    His Tag
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Anti-CD47 Mab (Human IgG4) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind Cynomolgus/Rhesus macaque CD47, His Tag (Cat. No. CD7-C52H1) with an affinity constant of 1.8 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
  • Gene ID:
    F7A802-1
  • Protein Synonyms:
    IAP|MER6|OA3|CD47
  • Protein/Peptide Name:
    CD47
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    15.8 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT
  • Cat. No.:
    103790-186

Specifications

About this item

Cynomolgus / Rhesus macaque CD47 Protein, His Tag, ACROBiosystems