Cynomolgus Monkey, Mouse Recombinant IL23A & IL12B (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Cynomolgus IL-23 alpha & Mouse IL-12 beta Heterodimer Protein, His Tag&Tag Free, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkeyMouse
- Tag sequence:His Tag&Tag Free
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Loaded Human IL-23 R, Fc Tag (Cat. No. ILR-H5254) on Protein A Biosensor, can bind Cynomolgus IL-23A & Mouse IL-12B Heterodimer Protein, His Tag&Tag Free (Cat. No. ILB-CM52W8) with an affinity constant of 9.4 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:A0A2K5TLV4-1 (IL23A) & P43432-1 (IL12B)
- Protein Synonyms:IL-23 alpha & IL-12 beta
- Protein/Peptide Name:IL23A & IL12B
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:20.3 kDa (IL23A) & 35.8 kDa (IL12B)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together