Dog Recombinant CD47 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Canine CD47 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Dog
- Tag sequence:His Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human SIRP alpha, Fc Tag (HPLC-verified) (Cat. No. SIA-H5251) at 5 μg/mL (100 μL/well) can bind Canine CD47, His Tag (Cat. No. CD7-C52H5) with a linear range of 5-78 ng/mL (QC tested).
- Gene ID:F1P6D7-1
- Protein Synonyms:IAP|MER6|OA3|CD47
- Protein/Peptide Name:CD47
- Purity:>90% as determined by reduced SDS-PAGE.
- Molecular Weight:15.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together