Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD24 (from HEK293 cells), Biotin
Human Recombinant CD24 (from HEK293 cells), Biotin
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant CD24 (from HEK293 cells), Biotin
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Biotin
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Fc, Avitag™
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human CD24, Fc,Avitag (Cat. No. CD4-H82F5) at 5 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Anti-CD24 MAb (SN3) with a linear range of 0.6-20 ng/mL (QC tested).
  • Gene ID:
    P25063-1
  • Protein Synonyms:
    CD24A|CD24
  • Protein/Peptide Name:
    CD24
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    31.4 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together