Advantage Economy Indirect Vent Goggles
Catalog # 470330-406
Supplier: DR INSTRUMENTS INC. SE
Specifications
- Frame color:Green
- Description:Advantage economy indirect vent goggles
- Cat. No.:470330-406
- Lens coating:Anti-fog
- Headband:18" adjustable elastic, green
- Frame material:PVC
- Lens material:PC
Specifications
About this item
Indirect vent and fog-free.
- Lightweight
- Green tinted
- Adjustable headband
Sellstrom advantage indirect vent economy protective goggles are lightweight, green tinted PVC. The soft body gently conforms to the face, providing a secure and comfortable fit. UV absorbing polycarbonate lens provide ultimate protection. Adjustable 18" green elastic headband secures the goggles comfortably to the face.
Certifications: Meets ANSI Z87.1-2003 standards. CUL certified to meet CAN/CSA Z94.3 standards.