Anti-Nucleocapsid Protein Hybridoma Polyclonal Antibody (AS61)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Type: Primary
Antigen: Nucleocapsid Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hybridoma
Isotype: Mouse IgG1 | Kappa
Reactivity:
Frequently Bought Together