Anti-Rituximab Hybridoma Polyclonal Antibody
Type: Primary
Antigen: Rituximab
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hybridoma
Isotype: IgG1/kappa
Reactivity:
Frequently Bought Together
Type: Primary
Antigen: Rituximab
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hybridoma
Isotype: IgG1/kappa
Reactivity: