Anti-Trastuzumab Hybridoma Polyclonal Antibody
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Type: Primary
Antigen: Trastuzumab
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hybridoma
Isotype: IgG1/kappa
Reactivity:
Frequently Bought Together