Order Entry
United States
ContactUsLinkComponent
Diversey™ Virex® Plus One-Step Disinfectant Cleaner and Deodorant, Essendant
Diversey™ Virex® Plus One-Step Disinfectant Cleaner and Deodorant, Essendant
Catalog # 76461-988
undefined
Diversey™ Virex® Plus One-Step Disinfectant Cleaner and Deodorant, Essendant
Catalog # 76461-988
Supplier Number:  DVO101102925

Specifications

  • Description:
    Virex® Plus One-Step Disinfectant Cleaner and Deodorant
  • Size:
    2× 1500 ml
  • Cat. No.:
    76461-988
  • Packaging:
    Plastic jug, closed-loop
  • Form:
    Liquid

Specifications

About this item

A one-step, quaternary-based disinfectant cleaner concentrate providing broad spectrum disinfection at 1:256 dilution. Use in healthcare and other facilities where cleaning and prevention of cross-contamination are critical. Bactericidal, virucidal and fungicidal. Kills MRSA and VRE. Meets bloodborne pathogen standards for decontaminating blood and body fluids.

  • One-step, quaternary-based disinfectant cleaner concentrate
  • Provides broad spectrum disinfection at 1:256 dilution
  • Bactericidal, virucidal and fungicidal
  • Kills MRSA and VRE
  • Blue in color with no added fragrance

Closed Loop—Designed for use with P&G Professional dispensing equipment.

Application: Disinfectant/Cleaner.

Applicable Material: Hard, Nonporous Surfaces.

Chemical Compound: Di-n-alkyl Dimethyl Ammonium Chloride.

Dirt Types: HIV-1; Influenza Type A; Pathogenic Bacteria; Trichophyton Mentagrophytes.