Order Entry
United States
ContactUsLinkComponent
Diversey™ End Bac® II Spray Disinfectant, Essendant
Diversey™ End Bac® II Spray Disinfectant, Essendant
Catalog # 76461-960
undefined
Diversey™ End Bac® II Spray Disinfectant, Essendant
Catalog # 76461-960
Supplier Number:  DVO04832

Specifications

  • Description:
    Diversey™ End Bac® II Spray Disinfectant
  • Size:
    12× 15 oz.
  • Cat. No.:
    76461-960
  • Packaging:
    Aerosol
  • Form:
    Liquid

Specifications

About this item

Disinfectant for use on pre-cleaned surfaces. Simply spray on hard surfaces then allow for air dry. Controls the growth of mold and mildew while eliminating odors at their source. Ideal for hospitals, locker rooms, shower stalls, floors and benches.

  • Disinfectant for use on pre-cleaned surfaces, unscented
  • Simply spray on hard surfaces then allow for air dry
  • Controls the growth of mold and mildew
  • Ideal for hospitals, locker rooms, shower stalls, floors and benches

Application: Disinfectant/Sanitizer.

Applicable Material: Ceramics; Enamel; Fiberglass; Glass; Laminates; Metal; Plastic; Vinyl.

Dirt Types: Bacteria; Fungus; Germs; Mildew; Viruses.