Diversey™ Divosan® Plus Broad Spectrum Sanitizer
76462-020
- Description:Divosan® Plus Broad Spectrum Sanitizer
- Size:1× 5 gal.
- Cat. No.:76462-020
- Packaging:Pail
Divosan® Plus is a peracetic acid based, broad spectrum sanitizer. It is suitable for use in dairies, breweries, soft drink and all other food processing operations. Divosan Plus is completely non-foaming and sanitizes effectively at temperatures as low as 40 °F.
- Completely non-foaming and suitable for clean-in-place (CIP) and spray applications
- Sanitizes and reduces bacteria in 1 minute, controls spoilage organisms; controls molds and yeast
- Effective at temperatures as low as 40 °F
Application: Beverage, Dairy and Food Processing Operations;
Applicable Material: Aluminum; Stainless Steel; Mild Steel; Brass; Copper; Galvanized Surfaces;
Chemical Compound: Acetic Acid; Hydrogen Peroxide; Peracetic Acid;
Dirt Types: Bacteria; Mold; Spoilage Organisms; Yeast.