Diversey™ Alpha-HP® Concentrated Multi-Surface Cleaner, Citrus Scent
Catalog # 76462-016
Supplier: Janitorial Supplies
Specifications
- Description:Alpha-HP® Concentrated Multi-Surface Cleaner, Citrus Scent
- Size:5000 ml
- Cat. No.:76462-016
- Packaging:Bottle, RTD®
- Form:Liquid
Specifications
About this item
All-in-one, multipurpose cleaner concentrate based on Accelerated Hydrogen Peroxide (AHP®) technology formulated with low levels of hydrogen peroxide. High productivity cleaning with no rinsing required. Cleans and brightens surfaces safely and easily.
- Accelerated Hydrogen Peroxide (AHP®) technology
- High productivity cleaning with no rinsing required
- Cleans and brightens surfaces safely and easily
- Three dilution rates provide a complete cleaning system
- Not for use on brass or copper
- Citrus scent
Three dilution rates provide a complete cleaning system for hard surfaces and carpet. Not for use on brass or copper.
Application: All-Purpose.
Applicable Material: Hard Non-Porous Surfaces.
Dirt Types: Dirt; Grease; Grime; Oil; Organic Matter; Smoke Film; Soap Scum.