Airfiltronix Ductless Fume Hood Filters, Cole-Parmer
Catalog # MSPP-EW3370536
Supplier: Cole Parmer
Specifications
- Description:Airfiltronix FRC-4 Formaldehyde Filter for Ductless Fume Hoods
- Cat. No.:MSPP-EW3370536
- Supplier no.:EW-33705-36
Specifications
About this item
These filters are compatible with the Aitfiltronix range of ductless fume hoods.
Ensure optimal performance of your ductless fume hood.