Order Entry
United States
ContactUsLinkComponent
Automatic Hands-Free Foam Hand Sanitizer/Soap Dispenser with Drip Tray, Alpine
Automatic Hands-Free Foam Hand Sanitizer/Soap Dispenser with Drip Tray, Alpine
Catalog # 76460-616
Supplier:  Avantor
undefined
Automatic Hands-Free Foam Hand Sanitizer/Soap Dispenser with Drip Tray, Alpine
Catalog # 76460-616
Supplier:  Avantor

Specifications

  • Capacity:
    1200 ml
  • Color:
    White
  • Description:
    ABS foam sanitizer dispenser
  • Cat. No.:
    76460-616
  • W×D×H:
    4.4×18"

Specifications

About this item

Dispenser works with most foam hand sanitizers or foam soap, making it ideal for use in offices, schools, warehouses, food service facilities, hospitals, and more.

  • For use with foam hand sanitizer and foam soap
  • Large 1200 ml capacity requires less frequent refills
  • Quiet, battery operated dispenser uses (4) C batteries (not included)
  • Lock included to prevent theft and vandalism
  • Automatic, hands-free operation for improved hand hygiene
  • Drip tray catches any foam so that floors remain clean

ADA compliant in force and one-handed operation, making it accessible to people with disabilities. Made from chemical, heat, and impact-resistant ABS plastic, these units are designed to provide long service life. Includes a drip tray to catch any foam so that floors remain clean. Wall mount design.