Order Entry
United States
ContactUsLinkComponent
Quick-DNA/RNA™ Blood Tube Kit, Zymo Research
Quick-DNA/RNA™ Blood Tube Kit, Zymo Research
Catalog # 76020-436
Supplier:  Zymo Research
CAS Number:  
undefined
Quick-DNA/RNA™ Blood Tube Kit, Zymo Research
Catalog # 76020-436
Supplier:  Zymo Research
Supplier Number:  R1151
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
    Total DNA
  • Sample type:
    Blood DNA/RNA collection tubes (e.g. PAXgene )
  • Target molecule:
    DNA
    RNA
  • Description:
    Quick-DNA/RNA Blood Tube Kit
  • Size:
    50 Prep
  • Environmentally Preferable:
  • Description:
    Quick-DNA/RNA Blood Tube Kit
  • Cat. No.:
    76020-436

Specifications

About this item

The Quick-DNA/RNA™ Blood Tube Kit is designed for use with the DNA/RNA Shield™ - Blood Collection Tube (R1150), enabling worry-free sample storage at ambient temperatures. The purification procedure uses Zymo-Spin column technology.

  • Superior Yields: Recover DNA and/or total RNA (including small/micro RNAs) without sample loss from the DNA/RNA Shield - Blood Collection Tubes (R1150).
  • Streamlined: No reagent removal (no pelleting) - Direct sample processing.
  • High-Quality: RNA is ready for all downstream applications including Next-Gen Sequencing, RT-PCR, etc. DNase I Included.

Simply bind the sample onto the Zymo-Spin™ IIICG Column with the aid of reservoirs (vacuum compatible). There is no reagent removal and no pelleting for easy direct whole tube processing. High quality DNA and/or total RNA from 3 ml whole blood is eluted into ≥50 µl of DNase/RNase-free water and is ready for any downstream application including RT-PCR, sequencing, etc

Technical Specifications
Equipment: Microcentrifuge, vortex, and vacuum/vacuum manifold
Purity: DNA and RNA is ready for Next-Gen sequencing, RTPCR, microarray, hybridization, etc. A260/A280, A260/A230: >1.8.
Sample Source: For use with DNA/RNA Shield™ - Blood Collection Tube (prefilled with 6 mL of DNA/RNA Shield™) for the direct collection of up to 3 ml whole-blood (human or animal).
Size Range: Genomic DNA ≥40 kb and Total RNA ≥17 nt
Yield: 50 µg DNA/RNA (binding capacity), ≥ 50 µl (elution volume)