ContactUsLinkComponent
Eisco Economy Microscope Slide Viewer
Eisco Economy Microscope Slide Viewer
Catalog # 470324-796
Supplier:  Wards
undefined
Eisco Economy Microscope Slide Viewer
Catalog # 470324-796
Supplier:  Wards
Supplier Number:  BI0276BV
Specifications
  • Description:
    Economy microscope slide viewer
  • Environmentally Preferable:
  • W×D×H:
    6.6×3.9×7.5"
  • Cat. No.:
    470324-796
Specifications
About this item
A fun introduction to the world of microscopy for students.
  • Lightweight and easy to use
  • Uses ambient light
  • Wireless, no battereies or light bulbs required
  • Compact design for easy storage and handling
This economical microscope slide viewer is light weight and economical. The unit does not require batteries or to be plugged into to an electrical source. Light from a lamp or natural light can be directed to the white surface and the light will reflect into the viewer.