Staphylococcus aureus V8 Nuclease S1
Catalog # RLMB-110-0010
Supplier: Rockland Immunochemical
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Species:Staphylococcus aureus V8
- Size:10000 units
- Enzyme Name:Nuclease S1
- Cat. No.:RLMB-110-0010
- Supplier no.:MB-110-0010
Specifications
About this item
All enzymes and related reagents are configured as an integrated system and have been thoroughly tested.