Specifications
- Description:Discovering qPCR kit
- No. of lab groups:4
- Cat. No.:470330-710
Specifications
About this item
Get hands-on experience with the principles and practice of qPCR.
- This kit must be used with a RT Thermal Cycler
- Learn to perform an advanced molecular biology procedure used in labs worldwide
- Quantify DNA concentration of 4 experimental samples using a standard curve
Quantitative PCR (qPCR or real-time PCR) simultaneously amplifies and detects targeted DNA allowing scientists to discover the starting amount of a specific DNA sequence in an experimental sample. This powerful and precise technology has become a cornerstone of modern genetics research, biological monitoring, and medical diagnostics.
For students, performing qPCR offers the challenge of performing a mathematically and technically advanced experiment, the opportunity to understand key molecular concepts through hands-on learning, and an essential skill for future research work.
In this specially adapted education qPCR experiment, students will quantify the DNA concentration of four experimental samples using a standard curve approach and then confirm the experiment’s specificity and accuracy through gel electrophoresis, melt curve analysis, and data analysis.
Time Required: qPCR Setup - 45 min; qPCR-90 min; Electrophoresis - 1 hr; Staining - 5 min to overnight, Analysis of results - 45 min.
Additional items needed but not included: qPCR Thermal Cycler, Microcentrifuge, 50 μl Micropipette, Electrophoresis equipment and light-box, Microwave or Hot Plate, Pipet Pump, Balance, Hot Gloves, Laboratory Gloves, Beaker, Distilled Water, Ice.