ContactUsLinkComponent
A-maize-ing Editing: Using CRISPR to Improve Crops
Catalog # 470330-680
Supplier:  Edvotek
undefined
A-maize-ing Editing: Using CRISPR to Improve Crops
Catalog # 470330-680
Supplier:  Edvotek
Supplier Number:  210
Specifications
  • Description:
    Using CRISPR kit
  • No. of lab groups:
    8
  • Cat. No.:
    470330-680
Specifications
About this item
Explore cutting edge biotechnology with this CRISPR simulation.
  • Hands-on CRISPR Simulation
  • Create Guide RNAs (gRNAs)
  • Use BLAST to verify gRNAs' specificity
  • Test your gRNAs using gel electrophoresis
  • Complete in approx. 1 hour 45 min
Students will assume the role of plant geneticists working to develop corn crops capable of surviving, and thriving, in a changing environment.
This experiment will allow students to develop an understanding of CRISPR-Cas9 applications in the laboratory, cleave DNA, and examine their results after gel electrophoresis.
Additional equipment needed but not included: DNA electrophoresis equipment, microwave or hot plate.
Ordering information: Kit will ship immediately upon ordering. Please call to place your order if you would like to specify a future date for shipment.
Caution: Some components require Freezer storage upon receipt.