Order Entry
United States
ContactUsLinkComponent
T4 DNA Ligase, Intact Genomics
T4 DNA Ligase, Intact Genomics
Catalog # 103753-180
CAS Number:  
undefined
T4 DNA Ligase, Intact Genomics
Catalog # 103753-180
Supplier Number:  3212
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    400 U/µl
  • Application:
  • Description:
    T4 DNA Ligase
  • Size:
    100000 Units
  • Environmentally Preferable:
  • Cat. No.:
    103753-180

Specifications

About this item

T4 DNA ligase displays significantly higher ligation efficiency than the nearest competitor.

  • 3 to 5X higher ligation efficiency than the nearest competitor
  • Extremely high purity
  • Useful for next-gen library preparation, cloning of PCR products, self-circularization of linear DNA and more.
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product.
  • The physical purity of this enzyme is ≥99% as assessed by SDS-PAGE with Coomassie® blue staining.
  • 10X T4 DNA ligase reaction buffer

T4 DNA ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA. This enzymes joins DNA fragments with either cohesive or blunt termini as well as repair single stranded nicks in duplex DNA, RNA or DNA/RNA hybrids.

Delivery: All our products are shipped on dry ice.