Specifications
- Subjects:General science
- Description:Bottle music
- No. of students:1
- Cat. No.:470330-046
- Grade:Elementary
Specifications
About this item
The nation's most exciting science competition now has kits for at-home use!
- Lesson plans and worksheets for two hands-on activities (lesson plans include extension suggestions for classroom or competition use)
- Materials to complete activities, excluding a few common household items (most materials may be reused)
- Educational standard: ESS-1
Elementary Science Olympiad Signature Events kits encourage hands-on learning and exploration for students in grades K-6, either at home or at school. The Bottle Music kit will help your child or student explore how a vibrating column of air creates a sound and how differing lengths of a column of air change the pitch of the sound. They will tap various lengths of 'Boomwhacker Tubes' to play simple songs.
All activities are aligned to Next Generation Science Standards, nationally recognized standards for science instruction. Parents, afterschool providers, and classroom teachers can successfully guide younger children through these activities, regardless of science background or knowledge. Older and more advanced children may be able to independently complete the activities with minimal assistance from adults.