ContactUsLinkComponent
exsig™ Mag Ultra-Sensitive Magnetic Bead Based Extraction System, Primerdesign
exsig™ Mag Ultra-Sensitive Magnetic Bead Based Extraction System, Primerdesign
Catalog # 76452-342
Supplier:  PRIMER DESIGN LTD
undefined
exsig™ Mag Ultra-Sensitive Magnetic Bead Based Extraction System, Primerdesign
Catalog # 76452-342
Supplier:  PRIMER DESIGN LTD
Supplier Number:  R35000
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    DNA/RNA extraction 5000 magnetic bead
  • Environmentally Preferable:
  • Cat. No.:
    76452-342
Specifications
About this item
exsig™ Mag facilitates a high-quality, high-throughput workflow solution by providing flexible extraction chemistry through its compatibility with widely used liquid handlers and extraction robots.
  • Ultra-sensitive nucleic acid purification
  • 5000 extractions
  • Fully automatable
  • Flexible chemistry compatible with a wide variety of sample types
  • Open platform that can be deployed on programmable extraction robotics
It compliments the genesig® real-time PCR COVID-19.