Order Entry
United States
ContactUsLinkComponent
Whatman™ Mini-UniPrep MultiCompressor, Whatman products (Cytiva)
Whatman™ Mini-UniPrep MultiCompressor, Whatman products (Cytiva)
Catalog # 76324-940
Supplier:  Cytiva
undefined
Whatman™ Mini-UniPrep MultiCompressor, Whatman products (Cytiva)
Catalog # 76324-940
Supplier:  Cytiva
Supplier Number:  MUPMCPBC8

Specifications

  • Capacity:
    8 plastic Mini-UniPrep vials ml
  • Description:
    Mini-UniPrep™ multi compressor with one blue 8 position tray
  • Cat. No.:
    76324-940
  • Width:
    324 mm

Specifications

About this item

Mini-UniPrep™ multicompressor including one 8-position tray for standard plastic Mini-UniPrep™ filter vials.

  • Streamlines syringeless filtration by filtering up to eight filter vials simultaneously
  • Designed for Cytiva's Mini-UniPrep™ standard plastic filter vial
  • Mini-UniPrep™ vials can be loaded directly into the autosampler

The newly redesigned Whatman™ Mini-UniPrep™ multicompressor streamlines filtration, saves time and reduces hand stress by enabling the filtration of eight Mini-UniPrep™ vials at once.

Ordering information: Additional trays are available, giving the added convenience of pre-loading up to eight Mini-UniPrep™ filter vials.
Delivery: The multiprocessor includes one blue 8-position tray.