Klenow Fragment (3'-5' exo-)
Klenow Fragment (3´→ 5´ exo-) is an N-terminal truncation of DNA Polymerase I which retains polymerase activity, but has lost the 5´→ 3´ exonuclease activity and has mutations (D355A, E357A) which abolish the 3´→ 5´ exonuclease activity.
- Enzyme Name:Klenow Fragment (3'-5' exo-)
- Enzyme Activity:5000 units/ml
Frequently Bought Together