Clinically relevant native human extracellular matrix (ECM) for cell culture coating.
- Native human extracellular matrix (ECM) for thin coating
- Clinically relevant and xeno-free for regenerative medicine applications
- Ready to use
- Supports various cell attachments
HumaMatrix Coat is isolated from nutrient-rich human tissue sourced strictly from tissue partners who comply with the requirements for transplantable human tissue. HumaMatrix Coat has undergone a proprietary manufacturing method to preserve native matrix proteins and growth factors. HumaMatrix Coat is supplied at 1 mg/ml for cell culture applications and can be diluted for custom applications.
HumaMatrix Coat addresses the growing need for more clinically relevant and animal-free human tissue constructs and models. Each lot of HumaMatrix Coat has undergone extensive testing to meet high safety and quality requirements for translational research needs.
All tissue has been tested for safety and found negative for communicable diseases and infectious agents.
- Storage Conditions:−20 °C
- Solubility:Soluble at pH ~2.0
- Application:Cell culture coating
- Concentration:1 mg/ml
- Formulation:Solution
- Tested Applications:Cell attachment