Order Entry
United States
ContactUsLinkComponent
HumaMatrix Native Human-Derived ECM, 20 mg/ml Solution, Humabiologics
HumaMatrix Native Human-Derived ECM, 20 mg/ml Solution, Humabiologics
Supplier:  HUMABIOLOGICS, INC
Certificates
undefined
HumaMatrix Native Human-Derived ECM, 20 mg/ml Solution, Humabiologics
Supplier:  HUMABIOLOGICS, INC

Specifications

  • Storage Conditions:
    −20 °C
  • Solubility:
    Soluble at pH ~2.0
  • Application:
    3D Hydrogel and Bioink
  • Concentration:
    20 mg/ml
  • Formulation:
    Solution
  • Tested Applications:
    Hydrogel formation and cell attachment

Specifications

Related Information

Frequently Bought Together