Clinically relevant native human extracellular matrix (ECM) for hydrogel and bioink applications.
- Native human extracellular matrix (ECM)
- Clinically relevant and xeno-free for regenerative medicine applications
- 3D hydrogel or bioink
- Ready to use
- Forms strong hydrogel
- Supports various cell attachments
HumaMatrix is isolated from nutrient-rich human tissue sourced strictly from tissue partners who comply with the requirements for transplantable human tissue. HumaMatrix has undergone a proprietary manufacturing method to preserve native matrix proteins and growth factors. HumaMatrix is supplied at a standard concentration of 10 mg/ml for strong hydrogel, bioprinting, and cell culture applications and can be diluted for custom applications.
HumaMatrix addresses the growing need for more clinically relevant and animal-free human tissue constructs and models. Each lot of HumaMatrix has undergone extensive testing to meet high safety and quality requirements for translational research needs.
All tissue has been tested for safety and found negative for communicable diseases and infectious agents.
- Storage Conditions:−20 °C
- Solubility:Soluble at pH ~2.0
- Application:3D Hydrogel and Bioink
- Concentration:10 mg/ml
- Formulation:Solution
- Tested Applications:Hydrogel formation and cell attachment