Order Entry
United States
ContactUsLinkComponent
Huma OsteoGelatin Medium Bloom Gelatin, Lyophilized, Humabiologics
Huma OsteoGelatin Medium Bloom Gelatin, Lyophilized, Humabiologics
Supplier:  HUMABIOLOGICS, INC
Certificates
undefined
Huma OsteoGelatin Medium Bloom Gelatin, Lyophilized, Humabiologics
Supplier:  HUMABIOLOGICS, INC

Specifications

  • Storage Conditions:
    15…30 °C
  • Solubility:
    Soluble in DI water
  • Application:
    Cell culture coating, 3D hydrogel, and bioink
  • Formulation:
    Lyophilized
  • Tested Applications:
    Hydrogel formation and cell attachment

Specifications

Related Information

Frequently Bought Together