Monarch® RNA Wash Buffer, New England Biolabs
103529-156
:
- Description:Monarch® RNA wash buffer
- Size:50 ml
- Cat. No.:103529-156
Need more RNA Wash Buffer? No problem. Monarch® kit components are all available for purchase separately.
- Can be used to wash bound RNA to remove contaminants including salts and enzymes
Monarch® RNA Wash Buffer is a component of the Monarch® Total RNA Miniprep Kit.
: Supplied as a 5X concentrate.