Monarch® RNA Priming Buffer, New England Biolabs
Catalog # 103529-154
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:Monarch® RNA priming buffer
- Size:56 ml
- Cat. No.:103529-154
Specifications
About this item
Need more RNA Priming Buffer? No problem. Monarch® kit components are all available for purchase separately.
- Used to wash bound RNA to remove contaminants including salts and enzymes
Monarch® RNA Priming Buffer is a component of the Monarch® Total RNA Miniprep Kit.
Delivery: Monarch® Priming Buffer is supplied in 2 bottles each containing 28 ml.