ContactUsLinkComponent
Monarch® RNA Purification Columns, New England Biolabs
Monarch® RNA Purification Columns, New England Biolabs
Catalog # 103529-146
CAS Number:  
Monarch® RNA Purification Columns, New England Biolabs
Catalog # 103529-146
Supplier Number:  T2007L
CAS Number:  
Specifications
  • Description:
    Monarch® RNA purification columns
  • Size:
    100 columns
  • Cat. No.:
    103529-146
Specifications
About this item
Need more RNA Purification Columns? No problem. NEB supplies all Monarch® kit components separately.
  • Are a component of the Monarch® Total RNA Miniprep Kit (103529-148)
  • Have a binding capacity of up to 100 µg of total RNA
  • Contain 100 columns and 100 collection tubes
  • Are manufactured using less plastic than other leading suppliers
Monarch® RNA Purification Columns, supplied with the Monarch® Total RNA Miniprep Kit, can purify up to 100 µg of total RNA from a variety of sample inputs.
Columns are supplied with Monarch® Collection Tubes II, and are compatible with vacuum manifolds.