Monarch® Total RNA Miniprep Enzyme Pack, New England Biolabs
Catalog # 103529-162
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total RNA
- Sample type:Plant samplesBacteriaBloodYeast/FungiCultured cellsHuman/animal cells and tissues
- Target molecule:RNA
- Description:Monarch® total RNA miniprep enzyme pack
- Size:1 pack
- Description:Monarch® total RNA miniprep enzyme pack
- Cat. No.:103529-162
Specifications
About this item
Need more enzymes for your Monarch® Total RNA Purification Kit? No problem. Monarch® Kit components are all available for purchase separately.
The Monarch® Total RNA Miniprep Enzyme Pack contains lyophilized Monarch® DNase I and Monarch® Proteinase K and their associated reaction buffers and resuspension buffers. The enzymes in this pack are optimized for use with the Monarch® Total RNA Miniprep Kit and are not recommended as substitutes for NEB's standard DNase I and Proteinase K.
Delivery information: The Monarch® Total RNA Miniprep Enzyme pack contains: Monarch® Proteinase K (20.8 mg), Monarch® Proteinase K Resuspension Buffer (1.5 ml), Monarch® Proteinase K Reaction Buffer (4 ml), Monarch® DNase I (275 units), Monarch® DNase I Reaction Buffer (4 ml) and Monarch® Nuclease-free Water (1.5 ml).