ContactUsLinkComponent
Monarch® gDNA Elution Buffer, New England Biolabs
  76339-784
 :  
Monarch® gDNA Elution Buffer, New England Biolabs
  76339-784
 :  T3016L
 :  
  • Description:
    Monarch® gDNA elution buffer
  • Size:
    34 ml
  • Cat. No.:
    76339-784
The Monarch® gDNA Elution Buffer a component of the Monarch® Genomic DNA Purification Kit (NEB #T3010), which can be used to purify up to 30 µg genomic DNA  from a variety of sample types.
Monarch® gDNA Elution Buffer is supplied in 10 mM Tris-HCl, pH 9.0, 0.1 mM EDTA and is suitable for long term storage of gDNA. Nucleases are inactivated both by the inclusion of EDTA and the pH of the solution.