About this item
Benefit from accurate and comprehensive analysis in low-volume lab setting. The DxH 500 hematology analyzer can help to improve clinical decision-making and efficiency. Built on proven cellular analysis technology, the DxH 500 provides precise platelet counts and accurate first-pass results. This means, can reduce costly test reruns and time-consuming manual reviews. The system’s intuitive interface also makes it easier to learn and operate, freeing the staff to focus on patient care.
- Spend time with the patients, not your analyzer
- Maximize uptime with built-in reliability, automatic start-up, efficient reagent usage and easy reagent replacement
- Free up time to focus on higher value activities with the ability to change all three reagents in less than five minutes
- Use 50% less reagent, consume reagents half as fast, and take less time changing and restocking reagents with the analyzer’s efficient approach
- Leverage single-sample precision
- Obtain accurate results with samples as small as 12 μl of blood (60 samples per hour)
- Save time by repeating testing on the same sample without another draw
Improve laboratory efficiency. Enable fast analysis with the analyzer’s unique fluidic design. Conserve valuable bench space with the DxH 500 hematology analyzer’s small footprint and integrated computer. Reduce operational complexity with the simplified user interface, which enables any command to be performed in three touches or less. Advance the efforts to go paperless with the analyzer’s bi-directional LIS Interface.
Test parameter: WBC, RBC, HGB, HCT, MCV, MCH, MCHC, RDW-SD, RDW-CV, PLT, MPV, LY%, LY#, MO%,MO#, NE%, NE#, EO%, EO#, BA%, BA# (menu/test parameters).
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76449-596
- 76449-594
- 76449-588
- 76449-592
- 76449-590
- 76449-584
- Sample Volume:
- 12 µl of venous or micro-collected whole blood or 20 µl of whole blood for pre-dilute analysis