ContactUsLinkComponent
VWR® Maximum Protection Sterile Duckbill Mask with Hood Loops
VWR® Maximum Protection Sterile Duckbill Mask with Hood Loops
Catalog # 76420-848
Supplier:  VWR International
undefined
VWR® Maximum Protection Sterile Duckbill Mask with Hood Loops
Catalog # 76420-848
Supplier:  VWR International
Specifications
  • Sterility:
    Sterile
  • Color:
    White
  • Description:
    Duckbill mask with hood loops
  • Cat. No.:
    76420-848
Specifications
About this item
The new sterile duckbill mask with hood loops expands upon the VWR® mask types in the popular cleanroom Maximum Protection line by providing a sterile option.
  • 3-ply masks, individually bagged
  • Sterile 10⁻⁶, gamma sterilization
  • Duckbill with hood loop configuration
  • Natural, latex-free
  • Low particle shedding
VWR® Maximum Protection sterile masks provide superior particle and bacterial filtration efficiency plus low particle shedding. These moisture-resistant cleanroom masks consist of three layers featuring a unique, lightweight inner layer of film for enhanced resistance.
Suitable for use in cleanroom environments, production, and research.
Packaging: Masks are packaged 1 per bag, 20 bags per pouch bag, 10 pouch bags per case.