Substage LED Illuminator for Boreal Beginner Microscope, Motic
Catalog # 470103-546
Supplier: VWR
Specifications
- Description:Substage illuminator
- Cat. No.:470103-546
- Supplier no.:55850-09
Specifications
About this item
Illuminator for use with the Boreal Beginner Microscope.
Add-on substage LED illuminator for cordless operation. LED element equal to 20 W tungsten bulb, powered by 3 AA rechargeable nickel metal hydride batteries. Each 8 hours recharge provides 4 to 50 hours of use. Recharger included, and plugs into any 115 V wall outlet.