High Performance FR Waffle Hoodies with Neck Gaiter, Lakeland Industries
Supplier: Lakeland
Certificates
About this item
Lakeland brings the incredibly popular waffle cut fabric to High Performance FR with permanent moisture wicking technology and some added stretch. The design ensures wearers will want this hoodie as part of the FR apparel layering system.
- Pullover design
- Integrated neck gaiter
- Waffle cut textured, inherently FR fabric
- Permanent moisture wicking properties
- Hand warmer pockets
Mid layers are the versatile garments that can help take the chill off for a few hours on a fall morning, or provide an extra layer of insulation for long, frigid winter days. Paired with a Base layer, Mid layers continue to pull moisture away from the body and move it to the outermost garment so it can dry. Lightweight, effective fabrics let Mid layers increase comfort without reducing mobility.
Certifications: NFPA 70E. NFPA 2112 - UL certified. APTV: 9.7 cal/cm². CAT: 2.
Specifications
- Catalog No:
- 76439-020
- 76439-022
- 76439-016
- 76439-038
- 76439-018
- 76439-024
- 76439-026
- 76439-028
- 76439-012
- 76439-034
- 76439-014
- 76439-036
- 76439-030
- 76439-010
- 76439-032
- Garment application:
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long