High Performance FR Long Sleeve Knit Crew, Black, Lakeland Industries
: Lakeland
Lightweight, Inherently FR Long Sleeve Knit Crew with permanent moisture wicking properties make this an extremely comfortable FR/AR base layer garment.
- Unparalleled CAT 2 protection at 16.4 cal/cm²
- Lightweight 6.95 oz. inherently FR fabric
- Excellent breathability
- Permanent moisture wicking properties
- Tag-free collar
- Drop shoulder design
- Left chest pocket
Base layers are the foundational key to the Lakeland FR/AR Layering System. To keep cool during hot summer days, and warm when its frigid cold outside, wearers need to be dry. Base layer shirts have the optimal 60/40 blend of hydrophobic and hydrophilic fibers that constantly pull moisture away from the body, move it to the outer surface of the garment, and allow it to dry. Staying dry is the key to keeping wearers comfortable and better protected.
: NFPA 70E. NFPA 2112 - UL certified. ATPV: 16.4 cal/cm². CAT: 2.
- Catalog No:
- 76438-908
- 76438-888
- 76438-884
- 76438-906
- 76438-886
- 76438-894
- 76438-890
- 76438-892
- 76438-904
- 76438-898
- 76438-900
- Garment application:
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Fabric:
- Fabric
- Fabric
- Fabric
- Fabric
- Fabric
- Fabric
- Fabric
- Fabric
- Fabric
- Fabric
- Fabric
- Weight:
- 6.95 oz.
- 6.95 oz.
- 6.95 oz.
- 6.95 oz.
- 6.95 oz.
- 6.95 oz.
- 6.95 oz.
- 6.95 oz.
- 6.95 oz.
- 6.95 oz.
- 6.95 oz.
Frequently Bought Together