mRNA Decapping
2000 units
Catalog # 76447-140
Supplier: New England Biolabs (NEB)
Supplier #: M0608S
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
mRNA Decapping Enzyme decaps m7G-capped mRNA producing a 5′ monophosphate end.
- Efficient replacement for Tobacco Acid Pyrophosphatase
- Cap0 and Cap1 are removed with equal efficiency
- Suitable for 5′ RLM-RACE and RNA-seq
- 1 µl will decap 20 µg of a 1.7 kb mRNA in 15 minutes
Specifications
Specifications
- Size:2000 units
- Cat. No.:76447-140
- Concentration:100000 units/ml
- Enzyme Name:mRNA Decapping