- Description:Disinfecting wipes, EPA registered
- Cat. No.:76443-228
- Packaging:300/container
- W×L:5.9×6.7"
EPA Registered disinfecting wipes kill 99.9% of bacteria and viruses, including coronavirus, with a contact time of 10 min.
- EPA Registered for range of viruses and bacteria, including coronavirus
- 10 min contact time for most viruses
Sanidry disinfecting wipes are intended for dry cleaning and sanitizing, and are formulated based on quats and alcohol according to the highest standards and regulations. The active substance in sanidry disinfecting wipes is considered by the US FDA as a 'No-Rinse' rinse free product GRAS (Generally Recognized as Safe) and is to be used as a rinse free product according to CFR 21 section 178.1010. It has been shown to be effective against viruses like influenza and coronavirus (CoV).
Wipes are recommended for cleaning where the use of water is not allowed, or for the replacement of rags, since rags represent a serious source of contamination. Recommended contact time for viruses is 10 min. Use Sanidry disinfecting wipes on the hard, nonporous surface of appliance exteriors, computer keyboards, bathroom fixtures, countertops, sinks, shower stalls, toilet seats, desks, doorknobs, floors, telephones, lab benches, and other hard nonporous surfaces made of acrylic, finished woodwork, glass, metal, plastic, and vinyl upholstery.