Order Entry
United States
ContactUsLinkComponent
tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
MOC:  Regulated
Certificates
undefined
tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
MOC:  Regulated
Certificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Identification C:
    Passes Test
  • USP - Alkalinity:
    Passes Test
  • USP - Water (H₂O):
    10.0 - 13.0 %
  • USP - Tartrate:
    Passes Test
  • USP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis):
    99.0 - 100.5 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Acidity or Alkalinity:
    Passes Test
  • EP/BP - Readily Carbonizable Substances:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤ 50 ppm
  • EP/BP - Oxalate (C₂O₄):
    ≤ 300 ppm
  • EP/BP - Sulfate (SO₄):
    ≤ 150 ppm
  • EP/BP - Water (H₂O):
    11.0 - 13.0 %
  • EP/BP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis):
    99.0 - 101.0 %
  • EP/BP - Endotoxin Concentration (EU/g):
    ≤ 2.5
  • JP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis):
    99.0 - 101.0 %
  • JP - pH (1 in 20):
    7.5 - 8.5
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤ 0.015 %
  • JP - Identification:
    Passes Test
  • JP - Sulfate (SO₄):
    ≤ 0.048 %
  • JP - Heavy Metals (as Pb):
    ≤ 10 ppm
  • JP - Arsenic (As):
    ≤ 2 ppm
  • JP - Tartrate:
    Passes Test
  • JP - Oxalate (C₂O₄):
    Passes Test
  • JP - Readily Carbonizable Substances:
    Passes Test
  • JP - Loss on Drying at 180°C:
    10.0 - 13.0 %
  • E. coli:
    Negative
  • P. aureginosa:
    Negative
  • ChP - Assay (dried basis):
    ≥ 99.0 %
  • ChP - Identification:
    Passes Test
  • ChP - Alkalinity:
    Passes Test
  • ChP - Clarity and Color of Solution:
    Passes Test
  • ChP - Chloride (Cl):
    ≤ 0.005 %
  • ChP - Sulfates (SO4):
    ≤ 0.015 %
  • ChP - Tartrate:
    Passes Test
  • ChP - Readily Carbonizable Substances:
    Passes Test
  • ChP - Loss on Drying, 180 °C:
    10.0 - 13.0 %
  • ChP - Calcium Salts or Oxalate:
    Passes Test
  • ChP - Iron (Fe):
    ≤ 0.001
  • ChP - Heavy Metals:
    ≤ 0.0005 %
  • ChP - Arsenic (As):
    ≤ 0.0001 %
  • ChP-Bacterial Endotoxin, EU/mg:
    ≤ 0.25

Specification Test Results

Related Information

Frequently Bought Together