ContactUsLinkComponent
Transport Swabs with Amies Gel, without Charcoal
Transport Swabs with Amies Gel, without Charcoal
Catalog # 76447-102
Supplier:  Hardy Diagnostics
undefined
Transport Swabs with Amies Gel, without Charcoal
Catalog # 76447-102
Supplier:  Hardy Diagnostics
Supplier Number:  4134BX
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Type:
    Dual swab
  • Description:
    Transporter swab with amies gel, without charcoal
  • Cat. No.:
    76447-102
  • Packaging:
    50/box
  • Tip style:
    Rayon tip
  • Application:
    Clinical
  • Cap color:
    Blue
Specifications
About this item
Transporter swab with amies gel, and without charcoal, features a dual plastic shaft, rayon swab tip, and blue cap.
  • Unique tube shape preserves gel media
  • Maximum gel for anaerobe preservation
  • Available with or without charcoal
  • Double plastic shaft
  • Regular rayon tip
Amies media is Stuart's original media formula modified by replacing the glycerophosphate in the original formula for an inorganic phosphate buffer. The metabolism of glycerophosphate by coliform organisms and other Gram negative rods, in Stuart's original formulation, resulted in proliferation of these organisms from throat, wound, and fecal specimens. Amies also found that NaCl at 0.3% w/v was optimal for the preservation of Neisseria gonorrhoeae. Calcium and magnesium salts were added in the belief that these ions were of importance in controlling the permeability of the bacterial cells and contributed to their survival.