Volatiles Mix D (High Level) for CLP Series Methods, SPEX CertiPrep
SXCLPV-D90H
:
- Concentration:2000 µg/ml
- Matrix:Methanol (P&T)
- Pack type:Ampoule, amber
- Description:Volatiles Mix D (High Level) for CLP Series Methods
- Size:1 ml
- Cat. No.:SXCLPV-D90H
Volatiles
CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.