To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Concentration:1000 µg/ml
- Matrix:Methanol
- Pack type:Ampoule, amber
- CAS No.:31262-37-0
- Description:Tetrahydrocannabivarin (THCV)
- Size:1 ml
- Description:Tetrahydrocannabivarin (THCV)
- Cat. No.:SXS-11057
Analytical standards for medicinal and recreational cannabis testing, DEA controlled substances.
Designed for Methods: State specific pesticide regulations:
- OAR 333-008-11
- HB 3460
- AOAC 2007-01
- EN 15662
While the legalization of cannabis for both medicinal and recreational purposes has been gaining speed, legislation and regulation has not necessarily kept pace. Out of a drive for self-regulation and significant consumer safety concerns, many producers and manufacturers are turning to testing labs in order to ensure that their products are of high quality and free of chemical contaminants.
SPEX CertiPrep offers ISO/IEC 17025 and ISO 17034 Certified Reference Materials (CRMs) for all of the common contaminants such as pesticide residues, residual solvents and heavy metals, as well as qualitative analysis CRMs, such as terpenes. As the industry demands change and regulations are put into place, we continually update our product offering.