Order Entry
United States
ContactUsLinkComponent
Tetrahydrocannabinolic acid (THCA) DEA Controlled Subatance, SPEX CertiPrep
Tetrahydrocannabinolic acid (THCA) DEA Controlled Subatance, SPEX CertiPrep
Catalog # SXS-11056
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Tetrahydrocannabinolic acid (THCA) DEA Controlled Subatance, SPEX CertiPrep
Catalog # SXS-11056
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  S-11056
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    Acetonitrile
  • Pack type:
    Ampoule, amber
  • CAS No.:
    23978-85-0
  • Description:
    Tetrahydrocannabinolic acid (THCA)
  • Size:
    1 ml
  • Description:
    Tetrahydrocannabinolic acid (THCA)
  • Cat. No.:
    SXS-11056

Specifications

About this item

Analytical standards for medicinal and recreational cannabis testing, DEA controlled substances.
Designed for Methods: State specific pesticide regulations:

  • OAR 333-008-11
  • HB 3460
  • AOAC 2007-01
  • EN 15662

While the legalization of cannabis for both medicinal and recreational purposes has been gaining speed, legislation and regulation has not necessarily kept pace. Out of a drive for self-regulation and significant consumer safety concerns, many producers and manufacturers are turning to testing labs in order to ensure that their products are of high quality and free of chemical contaminants.

SPEX CertiPrep offers ISO/IEC 17025 and ISO 17034 Certified Reference Materials (CRMs) for all of the common contaminants such as pesticide residues, residual solvents and heavy metals, as well as qualitative analysis CRMs, such as terpenes. As the industry demands change and regulations are put into place, we continually update our product offering.