Order Entry
United States
ContactUsLinkComponent
Decafluorotriphenylphosphine (DFTPP) for US EPA Methods 625 and 8270, SPEX CertiPrep
Decafluorotriphenylphosphine (DFTPP) for US EPA Methods 625 and 8270, SPEX CertiPrep
Catalog # SXECS-N-008
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Decafluorotriphenylphosphine (DFTPP) for US EPA Methods 625 and 8270, SPEX CertiPrep
Catalog # SXECS-N-008
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  ECS-N-008
CAS Number:  

Specifications

  • Concentration:
    50 µg/ml
  • Matrix:
    Methylene chloride
  • Pack type:
    Glass bottle, amber
  • CAS No.:
    5074-71-5
  • Description:
    Decafluorotriphenylphosphine (DFTPP)
  • Size:
    5.5 ml
  • Description:
    Decafluorotriphenylphosphine (DFTPP)
  • Cat. No.:
    SXECS-N-008

Specifications

About this item

ECS Standards for GC/MS

All ampulized standards are supplied with a pre-labeled transfer/storage vial. We want to make the transfer of ampulized standards as easy as possible. The label includes the catalog number, lot number, product description and expiration date.