Order Entry
United States
ContactUsLinkComponent
Polyethylene QC Standard Blank, SPEX CertiPrep
Polyethylene QC Standard Blank, SPEX CertiPrep
Catalog # SXCRM-PEBLK
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Polyethylene QC Standard Blank, SPEX CertiPrep
Catalog # SXCRM-PEBLK
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  CRM-PEBLK
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Pack type:
    Glass bottle, amber
  • Description:
    Polyethylene QC Standard Blank
  • Size:
    5 g
  • Cat. No.:
    SXCRM-PEBLK

Specifications

About this item

Consumer safety standards, phthalates in polyethylene QC standards.

Polyethylene (PE) is one of the world’s most common plastics. PE is used in a variety of common consumer products including children’s toys and care items. Current US regulations limit the concentrations of certain phthalates in childcare articles and children’s toys. Laboratories are tasked with the analysis of children’s toys for these potentially hazardous phthalates.